1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv
Solubility
Fully miscible in water.
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
RUO – Research Use Only
General References:
- Jung-Tang Wu.; Steve Lien-Chung Hsu.; Ming-Hsiu Tsai.; Weng-Sing Hwang. Inkjet Printing of Low-Temperature Cured Silver Patterns by Using AgNO3/1-Dimethylamino-2-propanol Inks on Polymer Substrates. J. Phys. Chem. 2011, 115 (22), 10940-10945.
- Salim Kadiwala.; Aravind V. Rayer.; Amr Henni. Kinetics of carbon dioxide (CO2) with ethylenediamine, 3-amino-1-propanol in methanol and ethanol, and with 1-dimethylamino-2-propanol and 3-dimethylamino-1-propanol in water using stopped-flow technique. Chemical Engineering Journal. 2012, 179 262-271.