Thermo Scientific Chemicals

1-Dimethylamino-2-propanol, 99%, Thermo Scientific Chemicals

Catalog number: B24234.30
250 g, Each
Thermo Scientific Chemicals

1-Dimethylamino-2-propanol, 99%, Thermo Scientific Chemicals

Catalog number: B24234.30
250 g, Each
Quantity
Catalog number: B24234.30
also known as B24234-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-16-7
IUPAC Name
1-(dimethylamino)propan-2-ol
Molecular Formula
C5H13NO
InChI Key
NCXUNZWLEYGQAH-UHFFFAOYNA-N
SMILES
CC(O)CN(C)C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.5%
Refractive Index
1.4180-1.4210 @ 20?C
Form
Liquid

Description

1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text