Thermo Scientific Chemicals

2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals

Catalog number: B24253.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dimethoxyphenol, 99%, Thermo Scientific Chemicals

Catalog number: B24253.14
25 g, Each
Quantity
Catalog number: B24253.14
also known as B24253-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-10-1
IUPAC Name
2,6-dimethoxyphenol
Molecular Formula
C8H10O3
InChI Key
KLIDCXVFHGNTTM-UHFFFAOYSA-N
SMILES
COC1=CC=CC(OC)=C1O
Solution Test
5% w/v in DMF: clear
Melting Point (clear melt)
52-58?C
Appearance (Color)
White or pale yellow to grey to brown
Form
Crystalline powder or crystals or fused solid
Assay (GC)
≥98.5%

Description

It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text