Thermo Scientific Chemicals

4-Chloromandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24264.06
5 g, Each
Thermo Scientific Chemicals

4-Chloromandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24264.06
5 g, Each
Quantity
Catalog number: B24264.06
also known as B24264-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
492-86-4
IUPAC Name
2-(4-chlorophenyl)-2-hydroxyacetic acid
Molecular Formula
C8H7ClO3
InChI Key
BWSFWXSSALIZAU-UHFFFAOYNA-N
SMILES
OC(C(O)=O)C1=CC=C(Cl)C=C1
Assay (Aqueous acid-base Titration)
>97.5 to <102.5%
Appearance (Color)
White to pale cream
Assay (Silylated GC)
>97.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text