Thermo Scientific Chemicals

2,6-Dichloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Catalog number: B24283.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dichloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Catalog number: B24283.14
25 g, Each
Quantity
Catalog number: B24283.14
also known as B24283-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99-30-9
IUPAC Name
2,6-dichloro-4-nitroaniline
Molecular Formula
C6H4Cl2N2O2
InChI Key
BIXZHMJUSMUDOQ-UHFFFAOYSA-N
SMILES
NC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥94.0%
Melting Point (clear melt)
183.5-194.5?C

Description

It is used as dyestuff, agricultural and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text