Thermo Scientific Chemicals

Ethyl (methylthio)acetate, 98%, Thermo Scientific Chemicals

Catalog number: B24285.14
25 g, Each
Thermo Scientific Chemicals

Ethyl (methylthio)acetate, 98%, Thermo Scientific Chemicals

Catalog number: B24285.14
25 g, Each
Quantity
Catalog number: B24285.14
also known as B24285-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4455-13-4
IUPAC Name
ethyl 2-(methylsulfanyl)acetate
Molecular Formula
C5H10O2S
InChI Key
MDIAKIHKBBNYHF-UHFFFAOYSA-N
SMILES
CCOC(=O)CSC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4570-1.4620 @ 20?C

Description

Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong alkalis.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text