Thermo Scientific Chemicals

Ethyl 3-(dimethylamino)acrylate, 99%, Thermo Scientific Chemicals

Catalog number: B24291.18
50 g, Each
Thermo Scientific Chemicals

Ethyl 3-(dimethylamino)acrylate, 99%, Thermo Scientific Chemicals

Catalog number: B24291.18
50 g, Each
Quantity
Catalog number: B24291.18
also known as B24291-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
924-99-2
IUPAC Name
ethyl (2Z)-3-(dimethylamino)prop-2-enoate
Molecular Formula
C7H13NO2
InChI Key
MVUMJYQUKKUOHO-WAYWQWQTSA-N
SMILES
CCOC(=O)C=C/N(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5105-1.5135 @ 20?C

Description

Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water 90 g/L (25°C).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text