Thermo Scientific Chemicals

Diaminomaleonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B24328.14
25 g, Each
Thermo Scientific Chemicals

Diaminomaleonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B24328.14
25 g, Each
Quantity
Catalog number: B24328.14
also known as B24328-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1187-42-4
IUPAC Name
(2Z)-diaminobut-2-enedinitrile
Molecular Formula
C4H4N4
InChI Key
DPZSNGJNFHWQDC-ARJAWSKDSA-N
SMILES
N\C(C#N)=C(/N)C#N
Appearance (Color)
Brown
Assay (UV/vis)
≥97.5%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text