3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24329.06
also known as B24329-06
Price (USD)/ Each
52.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
52.40
Add to cart
3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
Catalog numberB24329.06
Price (USD)/ Each
52.40
-
Add to cart
Chemical Identifiers
CAS18908-07-1
IUPAC Name1-isocyanato-3-methoxybenzene
Molecular FormulaC8H7NO2
InChI KeyNPOVTGVGOBJZPY-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.5420-1.5450 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only