Thermo Scientific Chemicals

5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals

Catalog number: B24336.03
1 g, Each
Thermo Scientific Chemicals

5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals

Catalog number: B24336.03
1 g, Each
Quantity
Catalog number: B24336.03
also known as B24336-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1544-85-0
IUPAC Name
2,2-difluoro-2H-1,3-benzodioxol-5-amine
Molecular Formula
C7H5F2NO2
InChI Key
CVYQRDKVWVBOFP-UHFFFAOYSA-N
SMILES
NC1=CC=C2OC(F)(F)OC2=C1
Refractive Index
1.4960-1.5010 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.0%
Form
Liquid

Description

5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text