Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab., Thermo Scientific Chemicals

Catalog number: B24340.14
25 g, Each
Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab., Thermo Scientific Chemicals

Catalog number: B24340.14
25 g, Each
Quantity
Catalog number: B24340.14
also known as B24340-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
45115-53-5
IUPAC Name
2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
Molecular Formula
C7H7F5O2
InChI Key
CLISWDZSTWQFNX-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCC(F)(F)C(F)(F)F
Form
Liquid
Appearance (Color)
Clear, colorless
Assay (GC)
≥96.0%
Refractive Index
1.3455-1.4505 @ 20?C
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text