Thermo Scientific Chemicals

2-(Trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24343.03
1 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24343.03
1 g, Each
Quantity
Catalog number: B24343.03
also known as B24343-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1423-27-4
IUPAC Name
[2-(trifluoromethyl)phenyl]boronic acid
Molecular Formula
C7H6BF3O2
InChI Key
JNSBEPKGFVENFS-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1C(F)(F)F
Appearance (Color)
White to pale brown
Assay (HPLC)
≥96.0%
Proton NMR
Conforms to structure
Assay (Aqueous acid-base Titration)
≥96.0%
Comment
Contains varying amounts of anhydride

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text