3,4-Diaminotoluene, 97%, Thermo Scientific Chemicals
3,4-Diaminotoluene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Diaminotoluene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B24378.36
also known as B24378-36
Price (USD)/ Each
143.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
143.00
Add to cart
3,4-Diaminotoluene, 97%, Thermo Scientific Chemicals
Catalog numberB24378.36
Price (USD)/ Each
143.00
-
Add to cart
Chemical Identifiers
CAS496-72-0
IUPAC Name4-methylbenzene-1,2-diamine
Molecular FormulaC7H10N2
InChI KeyDGRGLKZMKWPMOH-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)84.0-94.0?C
Appearance (Color)Pale brown to brown or purple
Assay (GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only