Ammonium cobalt(II) phosphate monohydrate, 98%, Thermo Scientific Chemicals
Ammonium cobalt(II) phosphate monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium cobalt(II) phosphate monohydrate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 g
Catalog number B24393.18
also known as B24393-18
Price (USD)/ Each
45.80
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
45.80
Add to cart
Ammonium cobalt(II) phosphate monohydrate, 98%, Thermo Scientific Chemicals
Catalog numberB24393.18
Price (USD)/ Each
45.80
-
Add to cart
Chemical Identifiers
CAS16827-96-6
IUPAC Nameλ²-cobalt(2+) ammonium hydrate phosphate
Molecular FormulaCoH6NO5P
InChI KeyPNSIYUITZYKGRY-UHFFFAOYSA-L
SMILES[NH4+].O.[Co++].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Heavy Metals (as Pb)≤25ppm
Appearance (Color)Pink to Purple
Elemental AnalysisCobalt, Co: ≥30.0 to ≤32.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only