Thermo Scientific Chemicals

4-Cyanostyrene, 97%, stab. with 0.05% 4-tert-butyl catechol, Thermo Scientific Chemicals

Catalog number: B24395.03
1 g, Each
Thermo Scientific Chemicals

4-Cyanostyrene, 97%, stab. with 0.05% 4-tert-butyl catechol, Thermo Scientific Chemicals

Catalog number: B24395.03
1 g, Each
Quantity
Catalog number: B24395.03
also known as B24395-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3435-51-6
Solution Test
(10% v/v in Diethyl ether) : Clear
Appearance (Color)
Clear, colorless
Assay (GC)
>96.0%
Solution Test
(10% v/v in Methanol) : Clear
Refractive Index
1.5750-1.5800 @ 20?C

Description

4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with polymerizing initiators, polymerization catalysts and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text