Thermo Scientific Chemicals

(+/-)-2-Amino-1-butanol, 97%, Thermo Scientific Chemicals

Catalog number: B24398.AE
100 mL, Each
Thermo Scientific Chemicals

(+/-)-2-Amino-1-butanol, 97%, Thermo Scientific Chemicals

Catalog number: B24398.AE
100 mL, Each
Quantity
Catalog number: B24398.AE
also known as B24398-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-20-8
IUPAC Name
2-aminobutan-1-ol
Molecular Formula
C4H11NO
InChI Key
JCBPETKZIGVZRE-UHFFFAOYNA-N
SMILES
CCC(N)CO
Form
Liquid or viscous liquid
Water Content (Karl Fischer Titration)
≤1.5%
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow to brown
Refractive Index
1.4500-1.4550 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text