Thermo Scientific Chemicals

(+/-)-2-Methylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B24405.22
100 g, Each
Thermo Scientific Chemicals

(+/-)-2-Methylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B24405.22
100 g, Each
Quantity
Catalog number: B24405.22
also known as B24405-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-07-9
IUPAC Name
2-methylpiperazine
Molecular Formula
C5H12N2
InChI Key
JOMNTHCQHJPVAZ-UHFFFAOYNA-N
SMILES
CC1CNCCN1
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White to yellow
Form
Crystalline powder or crystals and/or chunks
Melting Point (clear melt)
61-67°C
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text