3-Fluoro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
3-Fluoro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24411.03
also known as B24411-03
Price (USD)/ Each
64.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
64.90
Add to cart
3-Fluoro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Catalog numberB24411.03
Price (USD)/ Each
64.90
-
Add to cart
Chemical Identifiers
CAS69411-68-3
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)51.0-61.0?C
Assay (GC)≥96.0%
Appearance (Color)Pale cream to cream to brown
Water Content (Karl Fischer Titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only