Sodium hydroxide, pearl, 97%, Thermo Scientific Chemicals
Sodium hydroxide, pearl, 97%, Thermo Scientific Chemicals
Thermo Scientific™

Sodium hydroxide, pearl, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number B24414.36
also known as B24414-36
Price (USD)/ Each
31.65
Online exclusive
35.40 
Save 3.75 (11%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
31.65
Online exclusive
35.40 
Save 3.75 (11%)
Add to cart
Sodium hydroxide, pearl, 97%, Thermo Scientific Chemicals
Catalog numberB24414.36
Price (USD)/ Each
31.65
Online exclusive
35.40 
Save 3.75 (11%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or pellets
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

Solubility
Soluble in water, methanol, glycerol and ethanol. Insoluble in acetone and ether.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
RUO – Research Use Only

General References:

  1. Zhang, Y.; Zheng, S.; Fang, Z. Z.; Wang, X.; Du, H.; Zhang, Y. Reactive extraction of sodium hydroxide from alkali solutions for the separation of sodium and aluminum—Part II. Effect of diluent. Hydrometallurgy 2015, 156, 107-115.
  2. Lin, C. C.; Chu, C. R. Mass transfer performance of rotating packed beds with blade packings in carbon dioxide absorption into sodium hydroxide solution. Sep. Purif. Technol. 2015, 150, 196-203.