1,4-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
1,4-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dimethylpiperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number B24419.22
also known as B24419-22
Price (USD)/ Each
61.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
61.00
Add to cart
1,4-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
Catalog numberB24419.22
Price (USD)/ Each
61.00
-
Add to cart
Chemical Identifiers
CAS106-58-1
IUPAC Name1,4-dimethylpiperazine
Molecular FormulaC6H14N2
InChI KeyRXYPXQSKLGGKOL-UHFFFAOYSA-N
SMILESCN1CCN(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Refractive Index1.4450-1.4490 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only