Thermo Scientific Chemicals

2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: B24441.22
100 g, Each
Thermo Scientific Chemicals

2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: B24441.22
100 g, Each
Quantity
Catalog number: B24441.22
also known as B24441-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10312-55-7
IUPAC Name
2-aminobenzene-1,4-dicarboxylate
Molecular Formula
C8H5NO4
InChI Key
GPNNOCMCNFXRAO-UHFFFAOYSA-L
SMILES
NC1=CC(=CC=C1C([O-])=O)C([O-])=O
Appearance (Color)
Yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%
Form
Powder
Identification (FTIR)
Conforms

Description

2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text