Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
Solubility
Fully miscible in water.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
RUO – Research Use Only
General References:
- Sònia Abelló; Francisco Medina; Xavier Rodríguez; Yolanda Cesteros; Pilar Salagre; Jesús E. Sueiras; Didier Tichitb and Bernard Coq. Supported choline hydroxide (ionic liquid) as heterogeneous catalyst for aldol condensation reactions. Chem. Commun. 2004, (9), 1096-1097.
- Chunyan De; Bin Lu; He Lv; Yuanyuan Yu; Yu Bai; Qinghai Cai. One-Pot Synthesis of Dimethyl Carbonate from Methanol, Propylene Oxide and Carbon Dioxide Over Supported Choline hydroxide/MgO. Catalysis Letters. 2009, 128 (3), 459-464.