Thermo Scientific Chemicals

Potassium sulfamate, 98%, Thermo Scientific Chemicals

Catalog number: B24447.14
25 g, Each
Thermo Scientific Chemicals

Potassium sulfamate, 98%, Thermo Scientific Chemicals

Catalog number: B24447.14
25 g, Each
Quantity
Catalog number: B24447.14
also known as B24447-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13823-50-2
IUPAC Name
potassium sulfamate
Molecular Formula
H2KNO3S
InChI Key
BTAAXEFROUUDIL-UHFFFAOYSA-M
SMILES
[K+].NS([O-])(=O)=O
Appearance (Color)
White
Assay (ex Sulphated Ash)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid

Description

Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text