Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- A. S. Ermakov.; S. A. Serkov.; V. A. Tartakovskii.; T. S. Novikova.; L. I. Khmel'nitskii. Use of potassium sulfamate in the synthesis of heterocyclic nitramines.Chemistry of Heterocyclic Compounds. 1994, 30, (8),976-978 .