Potassium sulfamate, 98%, Thermo Scientific Chemicals
Potassium sulfamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium sulfamate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number B24447.36
also known as B24447-36
Price (USD)/ Each
424.65
Online exclusive
472.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
424.65
Online exclusive
472.00 
Save 47.35 (10%)
Add to cart
Potassium sulfamate, 98%, Thermo Scientific Chemicals
Catalog numberB24447.36
Price (USD)/ Each
424.65
Online exclusive
472.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13823-50-2
IUPAC Namepotassium sulfamate
Molecular FormulaH2KNO3S
InChI KeyBTAAXEFROUUDIL-UHFFFAOYSA-M
SMILES[K+].NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (ex Sulphated Ash)≥97.5%
FormCrystals or powder or crystalline powder or fused solid
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A. S. Ermakov.; S. A. Serkov.; V. A. Tartakovskii.; T. S. Novikova.; L. I. Khmel'nitskii. Use of potassium sulfamate in the synthesis of heterocyclic nitramines.Chemistry of Heterocyclic Compounds. 1994, 30, (8),976-978 .