Thermo Scientific Chemicals

2,6-Dichlorotoluene, 99%, Thermo Scientific Chemicals

Catalog number: B24463.22
100 g, Each
Thermo Scientific Chemicals

2,6-Dichlorotoluene, 99%, Thermo Scientific Chemicals

Catalog number: B24463.22
100 g, Each
Quantity
Catalog number: B24463.22
also known as B24463-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-69-4
IUPAC Name
1,3-dichloro-2-methylbenzene
Molecular Formula
C7H6Cl2
InChI Key
DMEDNTFWIHCBRK-UHFFFAOYSA-N
SMILES
CC1=C(Cl)C=CC=C1Cl
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5495-1.5525 @ 20?C
Assay (GC)
≥98.5%

Description

2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text