Thermo Scientific Chemicals

Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: B24468.22
100 g, Each
Thermo Scientific Chemicals

Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: B24468.22
100 g, Each
Quantity
Catalog number: B24468.22
also known as B24468-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27871-49-4
IUPAC Name
methyl (2S)-2-hydroxypropanoate
Molecular Formula
C4H8O3
InChI Key
LPEKGGXMPWTOCB-VKHMYHEASA-N
SMILES
COC(=O)[C@H](C)O
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Optical Rotation
-8.2 ± 0.2? (neat)
Identification (FTIR)
Conforms

Description

It is used in pharmaceutical industry, agricultural fields and is also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry, agricultural fields and is also used as a solvent.

Solubility
Miscible. Hydrolyzes in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text