Thermo Scientific Chemicals

Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals

Catalog number: B24470.06
5 g, Each
Thermo Scientific Chemicals

Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals

Catalog number: B24470.06
5 g, Each
Quantity
Catalog number: B24470.06
also known as B24470-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111865-47-5
IUPAC Name
benzyltrimethylazanium dibromine hydrobromide
Molecular Formula
C10H17Br3N
InChI Key
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
SMILES
Br.BrBr.C[N+](C)(C)CC1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
Orange
Melting Point (clear melt)
98.0-104.0?C
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text