Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals
Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B24470.14
also known as B24470-14
Price (USD)/ Each
233.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
233.00
Add to cart
Benzyltrimethylammonium tribromide, 98%, Thermo Scientific Chemicals
Catalog numberB24470.14
Price (USD)/ Each
233.00
-
Add to cart
Chemical Identifiers
CAS111865-47-5
IUPAC Namebenzyltrimethylazanium dibromine hydrobromide
Molecular FormulaC10H17Br3N
InChI KeyGSDDWZVWOXMLJZ-UHFFFAOYSA-N
SMILESBr.BrBr.C[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Orange
Melting Point (clear melt)98.0-104.0?C
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only