Isopropyl salicylate, 99%, Thermo Scientific Chemicals
Isopropyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl salicylate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24471.14
also known as B24471-14
Price (USD)/ Each
69.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
69.10
Add to cart
Isopropyl salicylate, 99%, Thermo Scientific Chemicals
Catalog numberB24471.14
Price (USD)/ Each
69.10
-
Add to cart
Chemical Identifiers
CAS607-85-2
IUPAC Namepropan-2-yl 2-hydroxybenzoate
Molecular FormulaC10H12O3
InChI KeyYEULQIJMIOWCHB-UHFFFAOYSA-N
SMILESCC(C)OC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5085-1.5115 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only