Thermo Scientific Chemicals

Isobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24472.36
500 g, Each
Thermo Scientific Chemicals

Isobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B24472.36
500 g, Each
Quantity
Catalog number: B24472.36
also known as B24472-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-30-1
IUPAC Name
2-methylpropanoyl chloride
Molecular Formula
C4H7ClO
InChI Key
DGMOBVGABMBZSB-UHFFFAOYSA-N
SMILES
CC(C)C(Cl)=O
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4055-1.4095 @ 20?C
Form
Liquid

Description

Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.

Solubility
Miscible with chloroform, glacial acetic acid, ether, toluene, methylene chloride and benzene.

Notes
Moisture sensitive. Incompatible with bases, water, strong oxidizing agents and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text