Thermo Scientific Chemicals

(S)-(+)-2,3-O-Isopropylideneglycerol, 98%, Thermo Scientific Chemicals

Catalog number: B24488.04
2 g, Each
Thermo Scientific Chemicals

(S)-(+)-2,3-O-Isopropylideneglycerol, 98%, Thermo Scientific Chemicals

Catalog number: B24488.04
2 g, Each
Quantity
Catalog number: B24488.04
also known as B24488-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22323-82-6
IUPAC Name
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Molecular Formula
C6H12O3
InChI Key
RNVYQYLELCKWAN-UHFFFAOYNA-N
SMILES
CC1(C)OCC(CO)O1
Form
Liquid
Optical Rotation
+11.5 ? 1? (c=5 in Methanol)
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text