Thermo Scientific Chemicals

3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B24497.06
5 g, Each
Thermo Scientific Chemicals

3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B24497.06
5 g, Each
Quantity
Catalog number: B24497.06
also known as B24497-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2039-86-3
IUPAC Name
1-bromo-3-ethenylbenzene
Molecular Formula
C8H7Br
InChI Key
KQJQPCJDKBKSLV-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(C=C)=C1
Assay (GC)
≥96.0%
Solution Test
10% in methanol: clear
Appearance (Color)
Clear colorless to pale yellow
Solution Test
10% in diethyl ether: clear
Stabilizer
0.1% 4-tert-butylcatechol

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text