2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals
2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24503.14
also known as B24503-14
Price (USD)/ Each
48.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
48.30
Add to cart
2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals
Catalog numberB24503.14
Price (USD)/ Each
48.30
-
Add to cart
Chemical Identifiers
CAS7623-09-8
IUPAC Name2-chloropropanoyl chloride
Molecular FormulaC3H4Cl2O
InChI KeyJEQDSBVHLKBEIZ-UHFFFAOYNA-N
SMILESCC(Cl)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥95.0%
Refractive Index1.4355-1.4420 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid

2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

Solubility
Reacts with water.

Notes
Store away from oxidizing agents, active metals, bases, water/ moisture.
RUO – Research Use Only

General References:

  1. Paulin N Wahjudi; Jin H Oh; Salam O Salman; Jason A Seabold; Damien C Rodger; Yu-Chong Tai; Mark E Thompson. Improvement of metal and tissue adhesion on surface-modified parylene C. Journal of Biomedical Materials Research, Part A. 2009, 89 (1), 206-214.
  2. Krzysztof Matyjaszewski; Jen-Lung Wang; Thomas Grimaud and Devon A. Shipp. Controlled/Living Atom Transfer Radical Polymerization of Methyl Methacrylate Using Various Initiation Systems. Macromolecules. 1998, 31 (5), 1527-1534.