2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals
2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B24509.22
also known as B24509-22
Price (USD)/ Each
48.20
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
48.20
Add to cart
2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals
Catalog numberB24509.22
Price (USD)/ Each
48.20
-
Add to cart
Chemical Identifiers
CAS115-70-8
IUPAC Name2-amino-2-ethylpropane-1,3-diol
Molecular FormulaC5H13NO2
InChI KeyIOAOAKDONABGPZ-UHFFFAOYSA-N
SMILESCCC(N)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Refractive Index1.4850-1.4900 @ 20?C
FormViscous liquid
Identification (FTIR)Conforms
It is used in pharmaceutical industry and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry and in medicine.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jung-Yeon Parka.; Sang Jun Yoon.; Huen Lee.; Ji-Ho Yoon.; Jae-Goo Shim.; Jae Keun Lee.; Byung-Yeon Min.; Hee-Moon Eum.; Min Cheol Kang. Solubility of carbon dioxide in aqueous solutions of 2-amino-2-ethyl-1,3-propanediol. Fluid Phase Equilib. 2002, 202 (2), 359-366 .
  2. Sang Jun Yoon .; Hyun-Song Lee.; Huen Lee.; Jeom-In Baek.; Ji-Ho Yoon. Hee-Moon Eum. Densities, Viscosities, and Surface Tensions of Aqueous 2-Amino-2-ethyl-1,3-propanediol Solutions.J. Chem. Eng. Data. 2002, 47 (1),30-32 .