Thermo Scientific Chemicals

2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B24509.36
500 g, Each
Thermo Scientific Chemicals

2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B24509.36
500 g, Each
Quantity
Catalog number: B24509.36
also known as B24509-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115-70-8
IUPAC Name
2-amino-2-ethylpropane-1,3-diol
Molecular Formula
C5H13NO2
InChI Key
IOAOAKDONABGPZ-UHFFFAOYSA-N
SMILES
CCC(N)(CO)CO
Appearance (Color)
Clear colorless to yellow
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Refractive Index
1.4850-1.4900 @ 20?C
Form
Viscous liquid
Identification (FTIR)
Conforms

Description

It is used in pharmaceutical industry and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry and in medicine.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text