Thermo Scientific Chemicals

Mucochloric acid, 99% (dry wt.), water <4.0%, Thermo Scientific Chemicals

Catalog number: B24523.36
500 g, Each
Thermo Scientific Chemicals

Mucochloric acid, 99% (dry wt.), water <4.0%, Thermo Scientific Chemicals

Catalog number: B24523.36
500 g, Each
Quantity
Catalog number: B24523.36
also known as B24523-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-56-9
IUPAC Name
(2Z)-2,3-dichloro-4-oxobut-2-enoic acid
Molecular Formula
C4H2Cl2O3
InChI Key
LUMLZKVIXLWTCI-IHWYPQMZSA-N
SMILES
OC(=O)C(Cl)=C(Cl)C=O
Appearance (Color)
White to cream
Form
Crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt basis)
Water Content (Karl Fischer Titration)
≤4.0%
Melting Point (clear melt)
124-130?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text