N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals
N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B24529.14
also known as B24529-14
Price (USD)/ Each
184.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
184.00
Add to cart
N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals
Catalog numberB24529.14
Price (USD)/ Each
184.00
-
Add to cart
Chemical Identifiers
CAS758-16-7
IUPAC NameN,N-dimethylmethanethioamide
Molecular FormulaC3H7NS
InChI KeySKECXRFZFFAANN-UHFFFAOYSA-N
SMILESCN(C)C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow or pale orange
FormLiquid
Assay (GC)≥96.0%
It is used in telecommunication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in telecommunication.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Horst Borrmann.; Ingmar Persson.; Magnus Sandström.; Christina M. V. Stålhandske. The crystal and liquid structures of N,N-dimethylthioformamide and N,N-dimethylformamide showing a stronger hydrogen bonding effect for C-H ⋯ S than of C-H ⋯ O.J. Chem. Soc., Perkin Trans. 2000, 2 ( 393-402), .
  2. Jan Näslund.; Ingmar Persson.; Magnus Sandström. Solvation of the Bismuth(III) Ion by Water, Dimethyl Sulfoxide, N,N‘-Dimethylpropyleneurea, and N,N-Dimethylthioformamide. An EXAFS, Large-Angle X-ray Scattering, and Crystallographic Structural Study. Inorg. Chem. 2000, 39 (18), 4012-4021 .