Thermo Scientific Chemicals

D-(-)-Asparagine monohydrate, 99%

Catalog number: B24556.14
25 g, Each
Thermo Scientific Chemicals

D-(-)-Asparagine monohydrate, 99%

Catalog number: B24556.14
25 g, Each
Quantity
Catalog number: B24556.14
also known as B24556-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5794-24-1
IUPAC Name
2-amino-3-carbamoylpropanoic acid hydrate
Molecular Formula
C4H10N2O4
InChI Key
RBMGJIZCEWRQES-UHFFFAOYNA-N
SMILES
O.NC(CC(N)=O)C(O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-31.0° ± 1.0° (c=5 in 5N HCl)

Description

D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (35 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text