Thermo Scientific Chemicals

Iron(II) gluconate dihydrate, 94%, Thermo Scientific Chemicals

Catalog number: B24561.36
500 g, Each
Thermo Scientific Chemicals

Iron(II) gluconate dihydrate, 94%, Thermo Scientific Chemicals

Catalog number: B24561.36
500 g, Each
Quantity
Catalog number: B24561.36
also known as B24561-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22830-45-1
IUPAC Name
λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Molecular Formula
C12H26FeO16
InChI Key
OKGNXSFAYMSVNN-SYAJEJNSSA-L
SMILES
O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Assay from Supplier's CofA
≥11.0% (% Fe)
Loss on Drying
<13.0%
Form
Powder
Identification (FTIR)
Conforms
Optical Rotation
+8? ±3? (c=1 in Water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text