Thermo Scientific Chemicals

Suberoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B24580.14
25 g, Each
Thermo Scientific Chemicals

Suberoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B24580.14
25 g, Each
Quantity
Catalog number: B24580.14
also known as B24580-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10027-07-3
IUPAC Name
octanedioyl dichloride
Molecular Formula
C8H12Cl2O2
InChI Key
PUIBKAHUQOOLSW-UHFFFAOYSA-N
SMILES
ClC(=O)CCCCCCC(Cl)=O
Assay (GC)
≥96.0%
Refractive Index
1.4665-1.4715 @ 20?C
Appearance (Color)
Clear colorless to brown
Form
Liquid

Description

Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text