Thermo Scientific Chemicals

2-(Trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B24591.06
5 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B24591.06
5 g, Each
Quantity
Catalog number: B24591.06
also known as B24591-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
447-61-0
IUPAC Name
2-(trifluoromethyl)benzaldehyde
Molecular Formula
C8H5F3O
InChI Key
ZDVRPKUWYQVVDX-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC=C1C=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4640-1.4680 @ 20°C

Description

2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

Solubility
Soluble in most organic solvents.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text