2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminopyrimidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B24594.36
also known as B24594-36
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
Catalog numberB24594.36
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS109-12-6
IUPAC Namepyrimidin-2-amine
Molecular FormulaC4H5N3
InChI KeyLJXQPZWIHJMPQQ-UHFFFAOYSA-N
SMILESNC1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.5%
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only