1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B24600.22
also known as B24600-22
Price (USD)/ Each
176.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
176.00
Add to cart
1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
Catalog numberB24600.22
Price (USD)/ Each
176.00
-
Add to cart
Chemical Identifiers
CAS6410-10-2
IUPAC Name(1Z)-1-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular FormulaC16H11N3O3
InChI KeyACHQDMGGRZPFAW-VLGSPTGOSA-N
SMILES[O-][N+](=O)C1=CC=C(NN=C2/C(=O)C=CC3=CC=CC=C23)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
lambda-max (UV-Vis)485-491 nm (in Toluene)
Solution Test0.1% (w/v) in Acetone : Clear
Appearance (Color)Red
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only