Thermo Scientific Chemicals

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals

Catalog number: B24601.06
5 g, Each
Thermo Scientific Chemicals

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals

Catalog number: B24601.06
5 g, Each
Quantity
Catalog number: B24601.06
also known as B24601-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1046-56-6
IUPAC Name
5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
Molecular Formula
C20H14N4
InChI Key
OTMYLOBWDNFTLO-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1
Melting Point (clear melt)
189.0-195.0?C
Appearance (Color)
Yellow
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms

Description

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text