Thermo Scientific Chemicals

2-Methyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Catalog number: B24612.22
100 g, Each
Thermo Scientific Chemicals

2-Methyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Catalog number: B24612.22
100 g, Each
Quantity
Catalog number: B24612.22
also known as B24612-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1120-64-5
IUPAC Name
2-methyl-4,5-dihydro-1,3-oxazole
Molecular Formula
C4H7NO
InChI Key
GUXJXWKCUUWCLX-UHFFFAOYSA-N
SMILES
CC1=NCCO1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4325-1.4355 @ 20°C

Description

2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N'-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N′-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

Solubility
Soluble in water (7051 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text