Thermo Scientific Chemicals

2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24613.06
5 g, Each
Thermo Scientific Chemicals

2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24613.06
5 g, Each
Quantity
Catalog number: B24613.06
also known as B24613-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100379-00-8
IUPAC Name
(2,6-dimethylphenyl)boronic acid
Molecular Formula
C8H11BO2
InChI Key
ZXDTWWZIHJEZOG-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=C1B(O)O
Appearance (Color)
White
Proton NMR
Conforms to structure
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder

Description

It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text