1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24620.06
also known as B24620-06
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
Add to cart
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
Catalog numberB24620.06
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS51851-37-7
IUPAC Nametriethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular FormulaC14H19F13O3Si
InChI KeyAVYKQOAMZCAHRG-UHFFFAOYSA-N
SMILESCCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3415-1.3465 @ 20?C
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.

Solubility
Miscible with alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only

General References:

  1. Kujawa, J.; Cerneaux, S.; Kujawski, W. Highly hydrophobic ceramic membranes applied to the removal of volatile organic compounds in pervaporation. Chem. Eng. J. 2015, 260, 43-54.
  2. Bahng, J. H.; Yeom, B.; Wang, Y.; Tung, S. O.; Hoff, J. D.; Kotov, N. Anomalous dispersions of/hedgehog/'particles. Nature 2015, 517 (7536), 596-599.