Thermo Scientific Chemicals

6-Fluoroindole, 98%, Thermo Scientific Chemicals

Catalog number: B24633.06
5 g, Each
Thermo Scientific Chemicals

6-Fluoroindole, 98%, Thermo Scientific Chemicals

Catalog number: B24633.06
5 g, Each
Quantity
Catalog number: B24633.06
also known as B24633-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
399-51-9
IUPAC Name
6-fluoro-1H-indole
Molecular Formula
C8H6FN
InChI Key
YYFFEPUCAKVRJX-UHFFFAOYSA-N
SMILES
FC1=CC=C2C=CNC2=C1
Melting Point (clear melt)
72.0-78.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Cream to pale brown
Assay (GC)
≥97.5%

Description

6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text