Thermo Scientific Chemicals

Ethyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B24658.0B
1000 g, Each
Thermo Scientific Chemicals

Ethyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B24658.0B
1000 g, Each
Quantity
Catalog number: B24658.0B
also known as B24658-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-40-0
IUPAC Name
ethyl 4-methylbenzene-1-sulfonate
Molecular Formula
C9H12O3S
InChI Key
VRZVPALEJCLXPR-UHFFFAOYSA-N
SMILES
CCOS(=O)(=O)C1=CC=C(C)C=C1
Form
Fused solid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
29.0-35.0?C

Description

Ethyl p-toluenesulfonate was used to develop an extraction method for methyl and ethyl esters of various sulfonic acids in active pharmaceutical ingredients using solid-phase micro extraction coupled to GC/MS in the selected ion monitoring mode. It was used in a study to develop a fast and accurate method for determination of residues of some common alkylating agents employed in drug synthesis by in situ derivatization-headspace-gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl p-toluenesulfonate was used to develop an extraction method for methyl and ethyl esters of various sulfonic acids in active pharmaceutical ingredients using solid-phase micro extraction coupled to GC/MS in the selected ion monitoring mode. It was used in a study to develop a fast and accurate method for determination of residues of some common alkylating agents employed in drug synthesis by in situ derivatization-headspace-gas chromatography-mass spectrometry.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text